Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate ≥95%

Supplier: AOBChem USA

Warning

82992-5G 82992-25G
77635-332EA 126 USD
77635-332 77660-408
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate ≥95%
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Ambient
MDL Number: MFCD06659589
CAS Number: 722497-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR