Tert-butyl 4-methyl-2-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

82961-5G 82961-1G 82961-500MG
77630-724EA 1766 USD
77630-724 77644-088 77652-070
Tert-butyl 4-methyl-2-(methylthio)benzoate ≥95%
Tert-butyl 4-methyl-2-(methylthio)benzoate

Precaución: For Research Use Only

Formula: 238.346 g/mol
MW: C₁₃H₁₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD34769392
CAS Number: 2918839-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR