5-Bromo-4-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

82887-5G 82887-1G 82887-25G 82887-500MG
77633-772EA 1093 USD
77633-772 77640-056 77656-954 77648-052
5-Bromo-4-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥95%
5-Bromo-4-methyl-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: For Research Use Only

Formula: 270.046 g/mol
MW: C₈H₇BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD28963192
CAS Number: 1598264-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR