Ethyl 2-(5-methoxy-2-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82731-1G 82731-5G 82731-500MG
77640-052EA 580 USD
77640-052 77656-950 77652-066
Ethyl 2-(5-methoxy-2-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-methoxy-2-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 222.237 g/mol
MW: C₁₂H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34769343
CAS Number: 1895203-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR