Ethyl 2-(2-methoxy-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82727-5G 82727-25G 82727-1G 82727-500MG
77630-720EA 1441 USD
77630-720 77636-224 77640-050 77648-048
Ethyl 2-(2-methoxy-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-methoxy-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 222.237 g/mol
MW: C₁₂H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34769340
CAS Number: 1895417-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR