Ethyl 2-(3-chloro-2,6-difluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82722-5G 82722-25G 82722-1G 82722-500MG
77630-718EA 1441 USD
77630-718 77636-222 77644-084 77652-064
Ethyl 2-(3-chloro-2,6-difluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-chloro-2,6-difluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₀H₇ClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34769336
CAS Number: 1897109-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR