Ethyl 2-(6-bromo-3-chloro-2-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82710-5G 82710-1G 82710-500MG
77656-948EA 2488 USD
77656-948 77644-082 77648-046
Ethyl 2-(6-bromo-3-chloro-2-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(6-bromo-3-chloro-2-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 309.516 g/mol
MW: C₁₀H₇BrClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34769330
CAS Number: 1897969-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR