Ethyl 2-(2-chloro-3,6-difluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82707-5G 82707-1G 82707-500MG
77630-716EA 1658 USD
77630-716 77640-048 77652-062
Ethyl 2-(2-chloro-3,6-difluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-3,6-difluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₀H₇ClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34769328
CAS Number: 1898158-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR