Ethyl 2-(2,6-difluoro-3-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82703-5G 82703-25G 82703-1G 82703-500MG
77630-714EA 1441 USD
77630-714 77636-220 77640-044 77653-096
Ethyl 2-(2,6-difluoro-3-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,6-difluoro-3-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 244.192 g/mol
MW: C₁₁H₁₀F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD34769326
CAS Number: 1898331-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR