Ethyl 2-(4-chloro-2-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82675-5G 82675-1G 82675-500MG
77630-712EA 1537 USD
77630-712 77640-040 77648-044
Ethyl 2-(4-chloro-2-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-chloro-2-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 242.656 g/mol
MW: C₁₁H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD11555531
CAS Number: 1256466-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR