Ethyl 2-(2,6-difluoro-4-methoxyphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82674-5G 82674-1G 82674-500MG
77630-710EA 1553 USD
77630-710 77644-080 77652-056
Ethyl 2-(2,6-difluoro-4-methoxyphenyl)-2-oxoacetate ≥97%
Ethyl 2-(2,6-difluoro-4-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 244.192 g/mol
MW: C₁₁H₁₀F₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16284970
CAS Number: 1272886-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR