Ethyl 2-(2-chloro-6-fluoro-3-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82671-5G 82671-1G 82671-500MG
77660-406EA 2245 USD
77660-406 77644-078 77648-042
Ethyl 2-(2-chloro-6-fluoro-3-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(2-chloro-6-fluoro-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 244.647 g/mol
MW: C₁₁H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD26681885
CAS Number: 1499035-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR