Ethyl 2-(2-chloro-5-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82625-5G 82625-1G 82625-500MG
77630-708EA 1586 USD
77630-708 77644-076 77652-054
Ethyl 2-(2-chloro-5-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-5-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 242.656 g/mol
MW: C₁₁H₁₁ClO₄
Storage Temperature: Ambient
MDL Number: MFCD26595206
CAS Number: 1502511-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR