Ethyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82617-5G 82617-1G 82617-500MG
77630-706EA 1586 USD
77630-706 77640-038 77652-052
Ethyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-chloro-5-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 280.628 g/mol
MW: C₁₁H₈ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28105019
CAS Number: 1891989-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR