Ethyl 2-(3-isopropylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82582-5G 82582-1G 82582-25G 82582-500MG
77633-768EA 1180 USD
77633-768 77640-036 77656-942 77648-038
Ethyl 2-(3-isopropylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-isopropylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 220.264 g/mol
MW: C₁₃H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11555681
CAS Number: 1256481-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR