Ethyl 2-(3-chloro-5-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82581-5G 82581-500MG 82581-1G
77630-702EA 1586 USD
77630-702 77653-094 77644-072
Ethyl 2-(3-chloro-5-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-chloro-5-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 226.656 g/mol
MW: C₁₁H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD11555534
CAS Number: 1256481-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR