Ethyl 2-(5-chloro-2-ethoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82542-5G 82542-1G 82542-500MG 82542-25G
77633-766EA 1310 USD
77633-766 77640-034 77653-090 77656-940
Ethyl 2-(5-chloro-2-ethoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-chloro-2-ethoxyphenyl)-2-oxoacetate
Formula: C₁₂H₁₃ClO₄
Storage Temperature: Ambient
MDL Number: MFCD14635321
CAS Number: 1368208-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR