Ethyl 2-(2-methyl-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82541-5G 82541-25G 82541-1G 82541-500MG
77633-764EA 1432 USD
77633-764 77660-404 77644-068 77652-050
Ethyl 2-(2-methyl-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-methyl-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 260.209 g/mol
MW: C₁₂H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD22688410
CAS Number: 1417458-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR