Ethyl 2-(3-fluoro-5-methoxyphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82523-5G 82523-1G 82523-25G 82523-500MG
77633-760EA 1432 USD
77633-760 77640-032 77656-938 77648-034
Ethyl 2-(3-fluoro-5-methoxyphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-fluoro-5-methoxyphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 226.201 g/mol
MW: C₁₁H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD11555654
CAS Number: 1256466-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR