Ethyl 2-(4-methoxy-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82512-5G 82512-25G 82512-1G 82512-500MG
77633-758EA 1432 USD
77633-758 77656-936 77644-064 77648-032
Ethyl 2-(4-methoxy-3-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-methoxy-3-(trifluoromethyl)phenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 276.209 g/mol
MW: C₁₂H₁₁F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD22688408
CAS Number: 1417511-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR