Ethyl-2-(thiomethyl)benzoylformate ≥95%

Supplier: AOBChem USA

Warning

82498-5G 82498-25G 82498-1G 82498-500MG
77633-756EA 932 USD
77633-756 77660-400 77644-062 77652-048
Ethyl-2-(thiomethyl)benzoylformate ≥95%
Ethyl-2-(thiomethyl)benzoylformate

Precaución: For Research Use Only

Formula: C₁₁H₁₂O₃S
MW: 224.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801441
CAS Number: 951888-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR