Ethyl 2-oxo-2-(3-(trifluoromethoxy)phenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

82494-5G 82494-1G 82494-25G 82494-500MG
77633-754EA 669 USD
77633-754 77640-030 77660-398 77648-030
Ethyl 2-oxo-2-(3-(trifluoromethoxy)phenyl)acetate ≥95%
Ethyl 2-oxo-2-(3-(trifluoromethoxy)phenyl)acetate

Precaución: For Research Use Only

Formula: 262.182 g/mol
MW: C₁₁H₉F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD16618265
CAS Number: 642447-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR