Ethyl-2-phenylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82486-5G 82486-500MG 82486-25G 82486-1G
77633-752EA 1026 USD
77633-752 77653-088 77656-934 77644-060
Ethyl-2-phenylbenzoylformate ≥95%
Ethyl-2-phenylbenzoylformate

Precaución: For Research Use Only

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801386
CAS Number: 947701-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR