Ethyl-4-methyl-1-naphthoylformate ≥95%

Supplier: AOBChem USA

Warning

82465-5G 82465-25G 82465-1G 82465-500MG
77633-748EA 1142 USD
77633-748 77656-930 77644-058 77648-028
Ethyl-4-methyl-1-naphthoylformate ≥95%
Ethyl-4-methyl-1-naphthoylformate

Precaución: For Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801435
CAS Number: 101093-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR