Ethyl-2,5-difluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82461-5G 82461-25G 82461-1G 82461-500MG
77633-746EA 932 USD
77633-746 77660-396 77644-056 77652-044
Ethyl-2,5-difluorobenzoylformate ≥95%
Ethyl-2,5-difluorobenzoylformate

Precaución: For Research Use Only

Formula: C₁₀H₈F₂O₃
MW: 214.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09965863
CAS Number: 1049131-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR