Ethyl 2-(2-bromo-6-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82451-5G 82451-25G 82451-1G 82451-500MG
77635-330EA 342 USD
77635-330 77660-394 77644-054 77648-026
Ethyl 2-(2-bromo-6-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-bromo-6-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₀H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD26523327
CAS Number: 1518047-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR