Ethyl-2,3-dichlorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82445-5G 82445-25G 82445-1G 82445-500MG
77633-744EA 1026 USD
77633-744 77660-392 77644-052 77652-042
Ethyl-2,3-dichlorobenzoylformate ≥95%
Ethyl-2,3-dichlorobenzoylformate

Precaución: For Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801402
CAS Number: 180868-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR