(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester ≥95%

Supplier: AOBChem USA

Warning

82440-5G 82440-25G 82440-1G 82440-500MG
77635-328EA 536 USD
77635-328 77656-928 77644-050 77652-040
(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester ≥95%
(2,4-Bis(trifluoromethylphenyl))oxoacetic acid ethyl ester

Precaución: For Research Use Only

Formula: C₁₂H₈F₆O₃
MW: 314.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01934869
CAS Number: 306936-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR