Ethyl-4-phenylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82426-5G 82426-1G 82426-25G 82426-500MG
77633-740EA 1232 USD
77633-740 77640-026 77656-924 77648-024
Ethyl-4-phenylbenzoylformate ≥95%
Ethyl-4-phenylbenzoylformate

Precaución: For Research Use Only

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801387
CAS Number: 6244-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR