Ethyl mesitylglyoxylate ≥95%

Supplier: AOBChem USA

Warning

82418-5G 82418-25G
77635-326EA 73 USD
77635-326 77660-388
Ethyl mesitylglyoxylate ≥95%
Ethyl mesitylglyoxylate

Precaución: For Research Use Only

Formula: C₁₃H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00151821
CAS Number: 5524-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR