Ethyl 2-(5-bromo-2-fluorophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82416-5G 82416-25G 82416-1G 82416-500MG
77635-324EA 432 USD
77635-324 77656-922 77644-046 77652-036
Ethyl 2-(5-bromo-2-fluorophenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-bromo-2-fluorophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 275.071 g/mol
MW: C₁₀H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD22375106
CAS Number: 668969-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR