Ethyl-3,5-difluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82401-5G 82401-25G
77635-322EA 114 USD
77635-322 77660-386
Ethyl-3,5-difluorobenzoylformate ≥95%
Ethyl 3,5-difluorobenzoylformate

Precaución: For Research Use Only

Formula: C₁₀H₈F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01319620
CAS Number: 208259-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR