Ethyl-4-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-fluorophenyl)-2-oxoacetate

82391-5G 82391-25G
77635-320EA 69 USD
77635-320 77656-920
Ethyl-4-fluorobenzoylformate ≥95%
Ethyl 4-fluorobenzoylformate

Precaución: For Research Use Only

Formula: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD01319617
CAS Number: 1813-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR