1-(4-Chloro-3,5-dimethylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

82379-5G 82379-1G 82379-25G 82379-500MG
77630-698EA 1441 USD
77630-698 77640-024 77660-384 77652-034
1-(4-Chloro-3,5-dimethylphenyl)cyclopentanol ≥95%
1-(4-Chloro-3,5-dimethylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 224.726 g/mol
MW: C₁₃H₁₇ClO
Storage Temperature: Ambient
MDL Number: MFCD34816853
CAS Number: 2921836-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR