2-(4-Fluoro-2-iodophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

82245-5G 82245-1G 82245-25G 82245-500MG
77630-696EA 1441 USD
77630-696 77640-022 77656-916 77652-032
2-(4-Fluoro-2-iodophenyl)-1,3-dioxolane ≥95%
2-(4-Fluoro-2-iodophenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 294.061 g/mol
MW: C₉H₈FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34769239
CAS Number: 1818236-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR