5-Bromo-2-chloro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

82208-5G 82208-1G 82208-500MG
77630-694EA 1788 USD
77630-694 77640-020 77652-030
5-Bromo-2-chloro-4-methoxybenzaldehyde ≥97%
5-Bromo-2-chloro-4-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD28536033
CAS Number: 1033723-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR