3-Iodo-2-(methylthio)benzo[b]thiophene ≥97%

Supplier: AOBChem USA

Warning

81644-5G 81644-25G 81644-1G 81644-500MG
77630-692EA 1441 USD
77630-692 77656-914 77644-044 77652-028
3-Iodo-2-(methylthio)benzo[b]thiophene ≥97%
3-Iodo-2-(methylthio)benzo[b]thiophene

Precaución: For Research Use Only

Formula: C₉H₇IS₂
Storage Temperature: Ambient
MDL Number: MFCD34816758
CAS Number: 1638536-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR