5-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

81302-5G 81302-1G 81302-500MG
77656-912EA 1988 USD
77656-912 77644-038 77652-022
5-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥95%
5-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: For Research Use Only

Formula: 307.979 g/mol
MW: C₁₃H₁₅BBrNO₂
Storage Temperature: Ambient
MDL Number: MFCD26404991
CAS Number: 1809946-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR