(2-Fluoro-4-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

81129-5G 81129-1G 81129-25G 81129-500MG
77633-738EA 1441 USD
77633-738 77640-014 77656-910 77648-018
(2-Fluoro-4-methoxyphenyl)(methyl)sulfane ≥95%
(2-Fluoro-4-methoxyphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 172.22 g/mol
MW: C₈H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD22489327
CAS Number: 66623-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR