N-Methoxy-N,4-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

80959-5G 80959-25G
77635-314EA 124 USD
77635-314 77660-372
N-Methoxy-N,4-dimethylbenzamide ≥95%
N-Methoxy-N,4-dimethylbenzamide

Precaución: For Research Use Only

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD02684311
CAS Number: 122334-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR