(5-Bromo-3-ethyl-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80956-5G 80956-25G 80956-1G 80956-500MG
77633-736EA 898 USD
77633-736 77656-908 77648-014 77648-016
(5-Bromo-3-ethyl-2-fluorophenyl)trimethylsilane ≥95%
(5-Bromo-3-ethyl-2-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₁H₁₆BrFSi
Storage Temperature: Ambient
MDL Number: MFCD34768540
CAS Number: 2918937-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR