Tert-butyl 2,3-difluoro-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

80627-5G 80627-25G 80627-1G 80627-500MG
77633-734EA 1055 USD
77633-734 77660-370 77644-036 77652-020
Tert-butyl 2,3-difluoro-4-methylbenzoate ≥95%
Tert-butyl 2,3-difluoro-4-methylbenzoate

Precaución: For Research Use Only

Formula: 228.235 g/mol
MW: C₁₂H₁₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22628078
CAS Number: 1203662-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR