(6-Bromo-2-chloro-3-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80478-5G 80478-25G 80478-1G 80478-500MG
77630-686EA 1441 USD
77630-686 77656-904 77644-034 77648-012
(6-Bromo-2-chloro-3-methylphenyl)trimethylsilane ≥95%
(6-Bromo-2-chloro-3-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₀H₁₄BrClSi
Storage Temperature: Ambient
MDL Number: MFCD34768340
CAS Number: 2918831-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR