(2-Chloro-6-fluoro-3-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80459-5G 80459-1G 80459-500MG
77660-368EA 2245 USD
77660-368 77644-032 77648-010
(2-Chloro-6-fluoro-3-methoxyphenyl)trimethylsilane ≥95%
(2-Chloro-6-fluoro-3-methoxyphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₀H₁₄ClFOSi
Storage Temperature: Ambient
MDL Number: MFCD17676532
CAS Number: 909543-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR