(3-Chloro-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80451-5G 80451-25G 80451-1G 80451-500MG
77633-732EA 1441 USD
77633-732 77660-366 77644-030 77648-008
(3-Chloro-2-fluorophenyl)trimethylsilane ≥95%
(3-Chloro-2-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 202.728 g/mol
MW: C₉H₁₂ClFSi
Storage Temperature: Ambient
MDL Number: MFCD34687578
CAS Number: 161957-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR