Tert-butyl 2,3-dichloro-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

80432-5G 80432-25G 80432-1G 80432-500MG
77630-682EA 1441 USD
77630-682 77636-212 77640-010 77652-016
Tert-butyl 2,3-dichloro-6-fluorobenzoate ≥95%
Tert-butyl 2,3-dichloro-6-fluorobenzoate

Precaución: For Research Use Only

Formula: 265.108 g/mol
MW: C₁₁H₁₁Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD34768311
CAS Number: 2918900-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR