5-(Pyridin-3-yl)thiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

80150-5G 80150-1G 80150-25G 80150-500MG
77635-312EA 425 USD
77635-312 77640-006 77660-362 77652-014
5-(Pyridin-3-yl)thiophene-2-carbaldehyde ≥97%
5-(Pyridin-3-yl)thiophene-2-carbaldehyde

Precaución: For Research Use Only

Formula: 189.234 g/mol
MW: C₁₀H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD00115001
CAS Number: 133531-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR