4-Bromo-5-chloro-1-methyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
80001-5G
80001-1G
80001-25G
80001-500MG
77633-726EA
1281
USD
77633-726
77640-002
77656-900
77648-004
4-Bromo-5-chloro-1-methyl-1H-pyrazole ≥97%
4-Bromo-5-chloro-1-methyl-1H-pyrazole
Precaución: For Research Use Only
Formula:
C₄H₄BrClN₂ MW: 195.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31414557 CAS Number: 1350323-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...