2,2,2-Trifluoro-1-(4-methoxynaphthalen-1-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

79845-5G 79845-1G 79845-25G 79845-500MG
77633-724EA 1378 USD
77633-724 77640-000 77656-898 77652-010
2,2,2-Trifluoro-1-(4-methoxynaphthalen-1-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-methoxynaphthalen-1-yl)ethanone

Precaución: For Research Use Only

Formula: C₁₃H₉F₃O₂
MW: 254.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380807
CAS Number: 169295-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR