1-(3-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

79536-5G 79536-25G 79536-1G 79536-500MG
77630-676EA 1441 USD
77630-676 77660-360 77644-024 77647-998
1-(3-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone ≥97%
1-(3-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 270.459 g/mol
MW: C₇H₃BrClF₂NO
Storage Temperature: Ambient
MDL Number: MFCD30717056
CAS Number: 2167281-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR